Proteintech
Manufacturer Code:159971AP150UL
Catalog # 50-173-0629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANT1/2 Fusion Protein Ag7787 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAC1 ADPATP carrier protein 1 ADP/ATP translocase 1 ANT ANT 1 ANT1/2 PEO2 PEO3 SLC25A4 T1; Adenine nucleotid translocator 2 fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotid translocator 2, fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocase-1; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); adenine nucleotide translocator 2 fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 2, fibroblast isoform (ATP-ADP carrier protein); ADP,ATP carrier protein 1; ADP,ATP carrier protein 2; ADP,ATP carrier protein, fibroblast isoform; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP translocase 1; ADP/ATP translocase 2; ADP/ATP translocase 2, N-terminally processed; ANT 1; ANT 2; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; Solute carrier family 25 member 4; Solute carrier family 25 member 5
Gene Aliases: 2F1; AAC1; AAC2; Anc1; ANT; ANT 1; ANT1; ANT2; AU019225; MTDPS12; PEO2; PEO3; PEOA2; SLC25A4; SLC25A5; T1; T2; T3
UniProt ID: (Human) P12235, (Human) P05141, (Rat) Q05962, (Mouse) P48962, (Rat) Q09073, (Mouse) P51881
Entrez Gene ID: (Human) 291, (Human) 292, (Rat) 85333, (Mouse) 11739, (Rat) 25176, (Mouse) 11740
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.