Invitrogen
Manufacturer Code:PA576637
Catalog # PIPA576637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ANT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAC1Adenine nucleotide translocator 1 (skeletal muscle)Adenine nucleotide translocator 1ADPADP ATP carrier protein 1ADP ATP carrier protein heart/skeletal muscle isoform T1ADP/ATP translocase 1ADT1&lowbarHUMANANT 1ANTANT1ATP carrier protein 1ATP carrier proteinheart/skeletal muscle isoform T1PEO2PEO3SLC25A4Solute carrier family 25 member 4T1; adenine nucleotide translocase-1; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP translocase 1; ANT 1; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; Solute carrier family 25 member 4
Gene Aliases: AAC1; Anc1; ANT; ANT 1; ANT1; AU019225; MTDPS12; PEO2; PEO3; PEOA2; SLC25A4; T1
UniProt ID: (Human) P12235, (Rat) Q05962, (Mouse) P48962
Entrez Gene ID: (Human) 291, (Rat) 85333, (Mouse) 11739
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.