Invitrogen
Manufacturer Code:MA527179
Catalog # PIMA527179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 26-153 of NPPA produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-atrial natriuretic peptide; Alpha-hANP; ANF; ANF 1-33; ANF 3-33; ANF 8-33; ANF ANF ANP ATFB6 Atrial natriuretic factor Atrial natriuretic peptide Atriopeptin Cardionatrin CDD-ANF CDP Natriuretic peptide A Natriuretic peptide precursor A Natriuretic peptides A NPPA PND Prepronatriodilatin Pronatriodilatin; ANP; Atrial natriuretic factor; Atrial natriuretic factor 1-33; Atrial natriuretic factor 3-33; Atrial natriuretic factor 8-33; Atrial natriuretic factor prohormone; Atrial natriuretic peptide; Atrial natriuretic peptide prohormone; Atriopeptigen; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; Auriculin-A; Auriculin-B; Auriculin-C; Auriculin-D; Cardiodilatin; cardiodilatin-related peptide; Cardionatrin; CDD; CDD 95-126; CDD-ANF; CDD-ANP (95-126); CDD-ANP (99-126); Kaliuretic peptide; KP; LANH; LANP; Long-acting natriuretic hormone; Long-acting natriuretic peptide; natriuretic peptide precursor A variant 1; Natriuretic peptides A; preproANP; preproCDD-ANF; prepronatriodilatin; Pro atrial natriuretic factor 1-30; Pro atrial natriuretic factor 31-67; Pro atrial natriuretic factor 79-98; Pro atrial natriuretic factor 99-126; Pro atrial natriuretic peptide 1-30; Pro atrial natriuretic peptide 31-67; Pro atrial natriuretic peptide 79-98; Pro atrial natriuretic peptide 95-126; proANF; proANF 1-30; proANF 31-67; proANF 79-98; proANF 99-126; proANP 1-30; proANP 31-67; proANP 79-98; proANP 95-126; URO; Urodilatin; Vessel dilator; VSDL
Gene Aliases: ANF; ANP; ATFB6; ATRST2; CDD; CDD-ANF; CDP; NPPA; PND
UniProt ID: (Human) P01160
Entrez Gene ID: (Human) 4878
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.