Invitrogen
Manufacturer Code:PA569330
Catalog # PIPA569330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ANO9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anoctamin-9; p53-induced gene 5 protein; PIG5 TMEM16J TP53I5 TM16J; Transmembrane protein 16J; tumor protein p53 inducible protein 5; Tumor protein p53-inducible protein 5
Gene Aliases: ANO9; PIG5; TMEM16J; TP53I5
UniProt ID: (Human) A1A5B4
Entrez Gene ID: (Human) 338440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.