Abnova
Manufacturer Code:PAB17333
Catalog # 89-118-989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human ANO9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ANO9.
For Research Use Only
Protein Aliases: Anoctamin-9; p53-induced gene 5 protein; PIG5; TMEM16J; TP53I5; Transmembrane protein 16J; tumor protein p53 inducible protein 5; Tumor protein p53-inducible protein 5
Gene Aliases: ANO9; PIG5; TMEM16J; TP53I5
UniProt ID: (Human) A1A5B4
Entrez Gene ID: (Human) 338440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.