Invitrogen
Manufacturer Code:PA526428
Catalog # PIPA526428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 106-135 amino acids from the N-terminal region of human ANO7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anoctamin-7; D-TMPP; D-TMPP DTMPP IPCA-5 IPCA5 NGEP PCANAP5 PCANAP5L TMEM16G Dresden-transmembrane protein of the prostate anoctamin-7 prostate cancer associated protein 5 transmembrane protein 16G; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; IPCA-5; New gene expressed in prostate; prostate cancer associated protein 5; Prostate cancer-associated protein 5; Transmembrane protein 16G
Gene Aliases: ANO7; D-TMPP; DTMPP; IPCA-5; IPCA5; NGEP; PCANAP5; PCANAP5L; TMEM16G
UniProt ID: (Human) Q6IWH7
Entrez Gene ID: (Human) 50636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.