Invitrogen
Manufacturer Code:PA5104532
Catalog # PIPA5104532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TM16G corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANO7 anoctamin 7 anoctamin-7 Dresden transmembrane protein of the prostate Dresden-transmembrane protein of the prostate DTMPP D-TMPP IPCA5 IPCA-5 New gene expressed in prostate NGEP PCANAP5 PCANAP5L prostate cancer associated protein 5 Prostate cancer-associated protein 5 TMEM16G Transmembrane protein 16G; Anoctamin-7; D-TMPP; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; IPCA-5; New gene expressed in prostate; prostate cancer associated protein 5; Prostate cancer-associated protein 5; Transmembrane protein 16G
Gene Aliases: ANO7; D-TMPP; DTMPP; IPCA-5; IPCA5; NGEP; PCANAP5; PCANAP5L; TMEM16G
UniProt ID: (Human) Q6IWH7
Entrez Gene ID: (Human) 50636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.