Invitrogen
Manufacturer Code:PA5111760
Catalog # PIPA5111760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 487-500 of human ANO6 (2nd extracellular loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900059G15Rik AA407480 Ano6 Anoctamin-6 AW554778 BDPLT7 F730003B03Rik SCAN channel SCTS Small-conductance calcium-activated nonselective cation channel TMEM16F Transmembrane protein 16F; Anoctamin-6; SCAN channel; Small-conductance calcium-activated nonselective cation channel; Transmembrane protein 16F
Gene Aliases: 2900059G15Rik; AA407480; ANO6; AW554778; BDPLT7; F730003B03Rik; SCTS; TMEM16F
UniProt ID: (Human) Q4KMQ2, (Mouse) Q6P9J9
Entrez Gene ID: (Human) 196527, (Rat) 315272, (Mouse) 105722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.