Proteintech
Manufacturer Code:207841AP150UL
Catalog # 50-173-3082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANO6 Fusion Protein Ag13937 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANO6 anoctamin 6 TMEM16F Transmembrane protein 16F; Anoctamin-6; SCAN channel; Small-conductance calcium-activated nonselective cation channel; Transmembrane protein 16F
Gene Aliases: 2900059G15Rik; AA407480; ANO6; AW554778; BDPLT7; F730003B03Rik; SCTS; TMEM16F
UniProt ID: (Human) Q4KMQ2, (Mouse) Q6P9J9
Entrez Gene ID: (Human) 196527, (Mouse) 105722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.