Invitrogen
Manufacturer Code:PA572157
Catalog # PIPA572157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 188-216 amino acids from the N-terminal region of human ANO4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anoctamin-4; Anoctamin-4 Transmembrane protein 16D ANO4 TMEM16D; Transmembrane protein 16D
Gene Aliases: ANO4; TMEM16D
UniProt ID: (Human) Q32M45
Entrez Gene ID: (Human) 121601
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.