Abnova
Manufacturer Code:H00257629K
Catalog # 89-207-310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ANKS4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ANKS4B peptide using ARM Technology.
For Research Use Only
Protein Aliases: Ankyrin repeat and SAM domain-containing protein 4B; FLJ38819 HARP MGC133380 MGC133381; Harmonin-interacting ankyrin repeat-containing protein; harmonin-interacting ankyrin-repeat containing protein; Harp
Gene Aliases: ANKS4B; HARP
UniProt ID: (Human) Q8N8V4
Entrez Gene ID: (Human) 257629
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.