Invitrogen
Manufacturer Code:PA545780
Catalog # PIPA545780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ANKS1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIDA-1; amyloid-beta precursor protein intracellular domain associated protein 1; Amyloid-beta protein intracellular domain-associated protein 1; ankyrin repeat and sterile alpha motif domain containing 1B AIDA AIDA-1 ANKS2 EB-1 EB1 MGC26087 cajalin-2; Ankyrin repeat and sterile alpha motif domain-containing protein 1B; cajalin 2; E2A-PBX1-associated protein; EB-1
Gene Aliases: AIDA; AIDA-1; AIDA-1b; ANKS1B; ANKS2; C030032C09Rik; cajalin-2; E530015N03Rik; EB-1; EB1; ENSMUSG00000078431; Gm10937; Gm1555; RGD1565556
UniProt ID: (Human) Q7Z6G8, (Mouse) Q8BIZ1, (Rat) P0C6S7
Entrez Gene ID: (Rabbit) 100338052, (Dog) 482621, (Horse) 100052606, (Human) 56899, (Mouse) 77531, (Rat) 314721
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.