Novus Biologicals
Manufacturer Code:NBP292519100UL
Catalog # NB026908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 261-510 of human ANKS1B (NP_858056.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIDA-1; AIDA-1AIDA amyloid-beta precursor protein intracellular domain associated protein 1 Amyloid-beta protein intracellular domain-associated protein 1 ANKS2 ankyrin repeat and sterile alpha motif domain containing 1B ankyrin repeat and sterile alpha motif domain-containing protein 1B cajalin 2 cajalin-2 E2A-PBX1-associated protein EB1 EB-1MGC26087; amyloid-beta precursor protein intracellular domain associated protein 1; Amyloid-beta protein intracellular domain-associated protein 1; Ankyrin repeat and sterile alpha motif domain-containing protein 1B; cajalin 2; E2A-PBX1-associated protein; EB-1
Gene Aliases: AIDA; AIDA-1; ANKS1B; ANKS2; cajalin-2; EB-1; EB1
UniProt ID: (Human) Q7Z6G8
Entrez Gene ID: (Human) 56899
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.