Proteintech
Manufacturer Code:247831AP150UL
Catalog # 50-173-4538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANKS1B Fusion Protein Ag20454 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIDA AIDA 1 ANKS1B ANKS2 cajalin 2 E2A PBX1 associated protein EB 1 EB1; AIDA-1; amyloid-beta precursor protein intracellular domain associated protein 1; Amyloid-beta protein intracellular domain-associated protein 1; Ankyrin repeat and sterile alpha motif domain-containing protein 1B; cajalin 2; E2A-PBX1-associated protein; EB-1
Gene Aliases: AIDA; AIDA-1; AIDA-1b; ANKS1B; ANKS2; C030032C09Rik; cajalin-2; E530015N03Rik; EB-1; EB1; ENSMUSG00000078431; Gm10937; Gm1555
UniProt ID: (Human) Q7Z6G8, (Mouse) Q8BIZ1
Entrez Gene ID: (Human) 56899, (Mouse) 77531
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.