Proteintech
Manufacturer Code:243331AP150UL
Catalog # 50-173-4304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANKRD6 Fusion Protein Ag19328 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKRD6 ankyrin repeat domain 6 DIVERSIN KIAA0957; Ankyrin repeat domain-containing protein 6; diego homolog; Diversin
Gene Aliases: ANKRD6; KIAA0957
UniProt ID: (Human) Q9Y2G4
Entrez Gene ID: (Human) 22881
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.