Invitrogen
Manufacturer Code:PA559301
Catalog # PIPA559301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ANKRD32 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat domain 32; Ankyrin repeat domain-containing protein 32; BRCT domain containing 1; BRCT domain-containing protein 1; BRCTD1 BRCTx BRCT domain containing 1 BRCT domain-containing protein 1 ankyrin repeat domain-containing protein 32; SMC5-SMC6 complex localization factor protein 1; Smc5/6 localization factor 1
Gene Aliases: ANKRD32; BRCTD1; BRCTx; SLF1
UniProt ID: (Human) Q9BQI6
Entrez Gene ID: (Human) 84250
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.