Novus Biologicals
Manufacturer Code:NBP185111
Catalog # NBP185111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:MALYDEDLLKNPFYLALQKCRPDLCSKVAQIHGIVLVPCKGSLSSSIQSTCQFESYILIPVEEHFQTLNGKDVFIQGNRIKLGAGFACLLSVPILFEETFYNEKEESFSILCI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat domain 27 (VPS9 domain); ankyrin repeat domain 27 (VPS9 domain) ankyrin repeat domain-containing protein 27 DKFZp434L0718 FLJ00040 VARP Vps9 domain and ankyrin-repeat-containing protein VPS9 domain-containing protein VPS9-ankyrin-repeat protein; Ankyrin repeat domain-containing protein 27; Vps9 domain and ankyrin-repeat-containing protein; VPS9 domain-containing protein; VPS9-ankyrin-repeat protein
Gene Aliases: ANKRD27; PP12899; VARP
UniProt ID: (Human) Q96NW4
Entrez Gene ID: (Human) 84079
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.