Invitrogen
Manufacturer Code:PA5109391
Catalog # PIPA5109391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human ANKRD2 (NP_001123453.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKRD2 Ankyrin repeat domain-containing protein 2 Arpp hArpp mArpp Skeletal muscle ankyrin repeat protein; ankyrin repeat domain 2 (stretch responsive muscle); Ankyrin repeat domain-containing protein 2; hArpp; mArpp; Skeletal muscle ankyrin repeat protein
Gene Aliases: ANKRD2; ARPP; mArpp
UniProt ID: (Human) Q9GZV1, (Mouse) Q9WV06
Entrez Gene ID: (Human) 26287, (Mouse) 56642, (Rat) 309374
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.