Novus Biologicals
Manufacturer Code:NB10086996
Catalog # NB10086996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1550 and 1600 of human ankyrin repeat domain 17 using the numbering given in entry NP_115593.3 (GeneID 26057). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat domain 17 ankyrin repeat domain-containing protein 17 Gene trap ankyrin repeat protein GTARSerologically defined breast cancer antigen NY-BR-16 KIAA0697FLJ22206 NY-BR-16; Ankyrin repeat domain-containing protein 17; Gene trap ankyrin repeat protein; Serologically defined breast cancer antigen NY-BR-16
Gene Aliases: ANKRD17; GTAR; KIAA0697; MASK2; NY-BR-16
UniProt ID: (Human) Q96I86
Entrez Gene ID: (Human) 26057
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.