Novus Biologicals
Manufacturer Code:NBP292635100UL
Catalog # NB027140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 900-1000 of human KANK1 (NP_055973.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKRD15ankyrin repeat domain 15 Ankyrin repeat domain-containing protein 15 KANKDKFZp451G231 KIAA0172MGC43128 Kidney ankyrin repeat-containing protein KN motif and ankyrin repeat domain-containing protein 1 KN motif and ankyrin repeat domains 1; Ankyrin repeat domain-containing protein 15; Kidney ankyrin repeat-containing protein; KN motif and ankyrin repeat domain-containing protein 1
Gene Aliases: ANKRD15; CPSQ2; KANK; KANK1; KIAA0172
UniProt ID: (Human) Q14678
Entrez Gene ID: (Human) 23189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.