Invitrogen
Manufacturer Code:PA543526
Catalog # PIPA543526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ANKH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANK; Ankylosis progressive homolog (mouse) ANK CCAL2 CMDJ CPPDD FLJ27166 HANK MANK; ankylosis, progressive homolog; Fn54 protein; progessive ankylosis-like protein; progressive ankylosis homolog; Progressive ankylosis protein; Progressive ankylosis protein homolog
Gene Aliases: ANK; ANKH; CCAL2; CMDJ; CPPDD; D15Ertd221e; HANK; KIAA1581; MANK; mKIAA1581; UNQ241/PRO274
UniProt ID: (Human) Q9HCJ1, (Mouse) Q9JHZ2, (Rat) P58366
Entrez Gene ID: (Human) 56172, (Horse) 100070193, (Dog) 491486, (Bovine) 511800, (Rabbit) 100343978, (Mouse) 11732, (Rat) 114506, (Guinea pig) 100719766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.