Novus Biologicals
Manufacturer Code:NBP257520
Catalog # NBP257520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ICTRGADMSVPDEKGNPPLWLALANNLEDIASTLVRHGCDATCWGPGPGGCLQTLLHRAIDENNEPTACFLIRSGCDVNSPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKHZN ankyrin repeat and FYVE domain containing 1 ankyrin repeat and FYVE domain-containing protein 1 ankyrin repeat hooked to zinc finger motif ankyrin repeats hooked to a zinc finger motif DKFZp686M19106 KIAA1255 Rabankyrin-5 ZFYVE14; Ankyrin repeat and FYVE domain-containing protein 1; ankyrin repeat hooked to zinc finger motif; Ankyrin repeats hooked to a zinc finger motif; Rabankyrin-5; Rank-5
Gene Aliases: ANKFY1; ANKHZN; BTBD23; KIAA1255; ZFYVE14
UniProt ID: (Human) Q9P2R3
Entrez Gene ID: (Human) 51479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.