Abnova
Manufacturer Code:MAB18244
Catalog # 89-338-169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S105-13 |
Immunogen | A synthetic peptide corresponding to amino acids 203-496 of human ANK2. |
Conjugate | ATTO565 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANK-2; ankyrin B; ankyrin, brain; Ankyrin-2; ankyrin-2, nonerythrocytic; Ankyrin-B; Brain ankyrin; DKFZp686M09125; DKFZp686P0948; FLJ38277; LQT4; Non-erythroid ankyrin
Gene Aliases: ANK-2; ANK2; ANKB; brank-2; LQT4
UniProt ID: (Human) Q01484
Entrez Gene ID: (Human) 287
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.