Invitrogen
Manufacturer Code:PA572812
Catalog # PIPA572812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANGPTL3 antibody for an 18 amino acid peptide near the carboxy terminus of human ANGPTL3. The immunogen is within the last 50 amino acids of ANGPTL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANG-5; angiopoietin 5; Angiopoietin-5; angiopoietin-like 3; Angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5 FHBL2 ANG-5 angiopoietin 5 angiopoietin-related protein 3; ANGPTL3(17-221); ANGPTL3(17-224); hypolipidemia
Gene Aliases: ANG-5; ANGPT5; ANGPTL3; ANL3; FHBL2; hypl; UNQ153/PRO179
UniProt ID: (Human) B2RCW1, (Mouse) Q9R182
Entrez Gene ID: (Human) 27329, (Mouse) 30924, (Rat) 502970
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.