Invitrogen
Manufacturer Code:PA547127
Catalog # PIPA547127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sf 21-derived recombit mouse Angiopoietin-like Protein 3/ANGPTL3 Ser17-Thr455 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5 FHBL2 ANG-5 angiopoietin 5 angiopoietin-related protein 3; ANGPTL3(17-224); hypolipidemia
Gene Aliases: Angptl3; hypl
UniProt ID: (Mouse) Q9R182
Entrez Gene ID: (Mouse) 30924
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.