Abnova
Manufacturer Code:PAB19841
Catalog # 89-121-373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids at C-terminus of human ANGPTL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ANGPTL3.
For Research Use Only
Protein Aliases: ANG-5; angiopoietin 5; Angiopoietin-5; angiopoietin-like 3; Angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3(17-221); ANGPTL3(17-224)
Gene Aliases: ANG-5; ANGPT5; ANGPTL3; ANL3; FHBL2; UNQ153/PRO179
UniProt ID: (Human) B2RCW1
Entrez Gene ID: (Human) 27329
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.