OriGene
Manufacturer Code:TA806289
Catalog # 50-168-1050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9E5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 24-491of human ANGPTL1 produced in SF9 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANG-3; ANG3 ANGPT3 ARP1 AngY UNQ162 dJ595C2.2; angioarrestin; angiopoietin 3; angiopoietin Y1; Angiopoietin-3; angiopoietin-like 1; Angiopoietin-like protein 1; Angiopoietin-related protein 1; dJ595C2.2 (angiopoietin Y1)
Gene Aliases: ANG3; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; PSEC0154; UNQ162; UNQ162/PRO188
UniProt ID: (Human) O95841
Entrez Gene ID: (Human) 9068
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.