Abnova
Manufacturer Code:PAB19789
Catalog # 89-121-317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids near C-terminus of human ANGPTL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ANGPTL1.
For Research Use Only
Protein Aliases: ANG-3; ANG3; angioarrestin; angiopoietin 3; angiopoietin Y1; Angiopoietin-3; angiopoietin-like 1; Angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); KIAA0351; UNQ162
Gene Aliases: ANG3; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; PSEC0154; UNQ162; UNQ162/PRO188
UniProt ID: (Human) O95841
Entrez Gene ID: (Human) 9068
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.