OriGene
Manufacturer Code:TA807191
Catalog # 50-168-1432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8C5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 20-320 of human ANGPT1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACONM HEL-S-284 ICRD OCA8 OPA9; ANG-1; Angiopoietin-1
Gene Aliases: AGP1; AGPT; ANG1; ANGPT1; KIAA0003
UniProt ID: (Human) Q15389
Entrez Gene ID: (Human) 284
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.