OriGene
Manufacturer Code:TA503407
Catalog # 50-167-4308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H8 |
Immunogen | Full length human recombit protein of human ANAPC2 produced in HEk293 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anaphase-promoting complex subunit 2; APC2; Cyclosome subunit 2; expressed during mesenchymal induction 4; induced during mesenchymal induction 4
Gene Aliases: 9230107K09Rik; AL024279; ANAPC2; APC2; Emi4; Imi4; KIAA1406; mKIAA1406
UniProt ID: (Human) Q9UJX6, (Mouse) Q8BZQ7
Entrez Gene ID: (Human) 29882, (Mouse) 99152, (Rat) 296558
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.