Novus Biologicals
Manufacturer Code:NBP271860PECY7
Catalog # N271860PEC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A6 |
Immunogen | Full length recombinant protein of human ANAPC2 (NP_037498) produced in HEk293 cell. |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: anaphase promoting complex subunit 2 anaphase-promoting complex subunit 2 APC2KIAA1406Cyclosome subunit 2 RP11-350O14.5; Anaphase-promoting complex subunit 2; APC2; Cyclosome subunit 2
Gene Aliases: ANAPC2; APC2; KIAA1406
UniProt ID: (Human) Q9UJX6
Entrez Gene ID: (Human) 29882
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.