Proteintech
Manufacturer Code:166641AP150UL
Catalog # 50-173-1096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AMZ2 Fusion Protein Ag10141 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMZ2 Archaemetzincin 2; Archaemetzincin-2; archaemetzincins-2; archeobacterial metalloproteinase-like 2; Archeobacterial metalloproteinase-like protein 2
Gene Aliases: AA408420; AMZ2; BM-014; ESTM12; RGD1304846; X83328
UniProt ID: (Human) Q86W34, (Rat) Q400C7, (Mouse) Q400C8
Entrez Gene ID: (Human) 51321, (Rat) 360650, (Mouse) 13929
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.