Invitrogen
Manufacturer Code:PA5109385
Catalog # PIPA5109385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 107-386 of human AMT (NP_001158184.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aminomethyltransferase (glycine cleavage system protein T); Aminomethyltransferase mitochondrial Amt EG434437 GCE GCST GCVT Glycine cleavage system T protein NKH; Aminomethyltransferase, mitochondrial; GCVT; Glycine cleavage system T protein
Gene Aliases: AMT; EG434437; GCE; GCST; GCVT; NKH
UniProt ID: (Human) P48728, (Mouse) Q8CFA2
Entrez Gene ID: (Human) 275, (Mouse) 434437, (Rat) 306586
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.