Invitrogen
Manufacturer Code:MA531869
Catalog # PIMA531869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C5D10 |
Immunogen | Purified recombit fragment of human PRKAG3 (AA: 9-151) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-AMP-activated protein kinase subunit gamma-3; 5'-AMP-activated protein kinase, gamma-3 subunit; AMPK gamma-3 chain; AMPK gamma3; AMPK subunit gamma-3; AMPKG3; protein kinase, AMP-activated, gamma 3 non-catalytic subunit
Gene Aliases: AMPKG3; PRKAG3
UniProt ID: (Human) Q9UGI9
Entrez Gene ID: (Human) 53632
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.