Invitrogen
Manufacturer Code:PA5113447
Catalog # PIPA5113447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human 5&prime-AMP-activated protein kinase subunit gamma-3 protein (1-210AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-AMP-activated protein kinase subunit gamma-3 5&prime-AMP-activated protein kinase gamma-3 subunit ampk gamma AMPK gamma3 AMPK gamma-3 chain AMPK subunit gamma-3 AMPKG3 OTTHUMP00000206393 PRKAG3 protein kinase AMP-activated non-catalytic subunit gamma 3 protein kinase AMP-activated gamma 3 non-catalytic subunit; 5'-AMP-activated protein kinase subunit gamma-3; 5'-AMP-activated protein kinase, gamma-3 subunit; AMPK gamma-3 chain; AMPK gamma3; AMPK subunit gamma-3; protein kinase, AMP-activated, gamma 3 non-catalytic subunit
Gene Aliases: AMPKG3; PRKAG3
UniProt ID: (Human) Q9UGI9
Entrez Gene ID: (Human) 53632
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.