Invitrogen
Manufacturer Code:PA522331
Catalog # PIPA522331
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 386 and 569 of AMPK gamma 2 (Uniprot IDNo.Q9UGJ0) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-AMP-activated protein kinase subunit gamma-2; 5-AMP-activated protein kinase, gamma-2 subunit; AAKG AAKG2 CMH6 H91620p WPWS 5&prime-AMP-activated protein kinase subunit gamma-2 AMPK subunit gamma-2; AMPK gamma-2 chain; AMPK gamma2; AMPK subunit gamma-2; H91620p; protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Gene Aliases: 2410051C13Rik; AAKG; AAKG2; AI854673; CMH6; H91620p; PRKAG2; WPWS
UniProt ID: (Human) Q9UGJ0, (Mouse) Q91WG5
Entrez Gene ID: (Human) 51422, (Mouse) 108099
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.