Invitrogen
Manufacturer Code:PA513795
Catalog # PIPA513795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 265-295 amino acids from the central region of human AMPK gamma (PRKAG1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-AMP-activated protein kinase gamma-1 subunit AMPK gamma-1 chain AMPKg; 5'-AMP-activated protein kinase subunit gamma-1; 5'-AMP-activated protein kinase, gamma-1 subunit; AMPK gamma-1 chain; AMPK gamma1; AMPK subunit gamma-1; AMPKg; protein kinase, AMP-activated, gamma 1 non-catalytic subunit
Gene Aliases: AA571379; AMPKG; BB036179; Prkaac; PRKAG1
UniProt ID: (Human) P54619, (Mouse) O54950
Entrez Gene ID: (Human) 5571, (Mouse) 19082
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.