Invitrogen
Manufacturer Code:PA5112885
Catalog # PIPA5112885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human 5&prime-AMP-activated protein kinase subunit beta-2 protein (1-272AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-AMP-activated protein kinase subunit beta-2; 5'-AMP-activated protein kinase, beta-2 subunit; 5730553K21Rik 5&prime-AMP-activated protein kinase subunit beta-2 5&prime-AMP-activated protein kinase beta-2 subunit AMP-activated protein kinase beta 2 non-catalytic subunit AMP-activated protein kinase beta-2 regulatory subunit AMPK beta 2 AMPK beta-2 chain AMPK subunit beta-2 AMPKbeta2 AW049591 BB124140 Prkab2 protein kinase AMP-activated non-catalytic subunit beta 2 protein kinase AMP-activated beta 2 non-catalytic subunit; AMP-activated protein kinase beta 2 non-catalytic subunit; AMP-activated protein kinase beta-2 regulatory subunit; AMPK beta 2; AMPK beta-2 chain; AMPK subunit beta-2; protein kinase, AMP-activated, beta 2 non-catalytic subunit
Gene Aliases: 5730553K21Rik; AW049591; BB124140; PRKAB2
UniProt ID: (Human) O43741, (Rat) Q9QZH4, (Mouse) Q6PAM0
Entrez Gene ID: (Human) 5565, (Rat) 64562, (Mouse) 108097
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.