Invitrogen
Manufacturer Code:MA537717
Catalog # PIMA537717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1420CT832.86.25 |
Immunogen | recombinant of human PRKAB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1300015D22Rik 5'-AMP-activated protein kinase 40 kDa subunit 5-AMP-activated protein kinase beta subunit 5'-AMP-activated protein kinase beta-1 subunit 5'-AMP-activated protein kinase subunit beta-1 5'-AMP-activated protein kinase subunit beta-1-like protein 5'-AMP-activated protein kinase beta subunit AAKB1 AMP-activated protein kinase beta subunit AMPK AMPK beta 1 AMPK beta -1 chain AMPK beta-1 chain AMPK subunit beta-1 AMPKb AMPK-beta1 AU021155 E430008F22 HAMPKb kinase AMPK-beta1 kinase pAMPK pAMPK PRKAB1 protein kinase AMP-activated non-catalytic subunit beta 1 protein kinase AMP-activated beta 1 non-catalytic subunit protein kinase AMP-activated noncatalytic beta-1; 5'-AMP-activated protein kinase 40 kDa subunit; 5'-AMP-activated protein kinase beta-1 subunit; 5'-AMP-activated protein kinase subunit beta-1; 5'-AMP-activated protein kinase, beta subunit; 5-AMP-activated protein kinase beta subunit; AMP-activated protein kinase beta subunit; AMPK beta -1 chain; AMPK beta 1; AMPK beta-1 chain; AMPK subunit beta-1; AMPKb; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1
Gene Aliases: 1300015D22Rik; AMPK; AU021155; E430008F22; HAMPKb; PRKAB1
UniProt ID: (Human) Q9Y478, (Mouse) Q9R078, (Rat) P80386
Entrez Gene ID: (Human) 5564, (Mouse) 19079, (Rat) 83803
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.