Novus Biologicals
Manufacturer Code:NBP303523100UL
Catalog # NB092571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human AMPK beta 1 (NP_006244.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-AMP-activated protein kinase beta-1 subunit; 5'-AMP-activated protein kinase beta-1 subunit AMP-activated protein kinase beta subunit AMPK AMPK beta -1 chain AMPK beta 15'-AMP-activated protein kinase subunit beta-1 AMPK subunit beta-1 AMPKb HAMPKb MGC17785 protein kinase AMP-activated beta 1 non-catalytic subunit protein kinase AMP-activated noncatalytic beta-1; 5'-AMP-activated protein kinase subunit beta-1; AMP-activated protein kinase beta subunit; AMPK beta -1 chain; AMPK beta 1; AMPK subunit beta-1; AMPKb; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1
Gene Aliases: AMPK; HAMPKb; PRKAB1
UniProt ID: (Human) Q9Y478
Entrez Gene ID: (Human) 5564
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.