Invitrogen
Manufacturer Code:PA519367
Catalog # PIPA519367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CPLDALNTTKP) corresponding to the internal amino acids of PRKAA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK-alpha-2 chain; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; prkaa2; PRKAA2 protein kinase AMP-activated alpha 2 catalytic subunit AMPK AMPK2 PRKAA 5&prime-AMP-activated protein kinase catalytic alpha-2 chain AMP-activated protein kinase alpha 2 catalytic subunit AMPK-alpha-2 chain; protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Aliases: 2310008I11Rik; A830082D05; AMPK; AMPK2; AMPKa2; AMPKalpha2; PRKAA; PRKAA2
UniProt ID: (Human) P54646, (Rat) Q09137, (Mouse) Q8BRK8
Entrez Gene ID: (Human) 5563, (Rat) 78975, (Mouse) 108079
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.