Invitrogen
Manufacturer Code:PA5114773
Catalog # PIPA5114773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRKAA2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310008I11Rik 5'-AMP-activated protein kinase catalytic subunit alpha-2 5'-AMP-activated protein kinase catalytic alpha-2 chain A830082D05 ACACA kinase acetyl-CoA carboxylase kinase AMP-activated protein kinase AMP-activated protein kinase alpha-2 subunit variant 2 AMP-activated protein kinase alpha-2 subunit variant 3 AMP-activated protein kinase alpha-2 variant B Ampk AMPK alpha 2 AMPK alpha-2 chain AMPK subunit alpha-2 AMPK2 AMPKa2 AMPKalpha2 AMPK-alpha-2 chain EC 2.7.11.1 HMGCR kinase Hydroxymethylglutaryl-CoA reductase kinase PRKAA prkaa2 protein kinase AMP-activated catalytic subunit alpha 2 protein kinase AMP-activated alpha 2 catalytic subunit; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK-alpha-2 chain; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; prkaa2; protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Aliases: 2310008I11Rik; A830082D05; AMPK; AMPK2; AMPKa2; AMPKalpha2; PRKAA; PRKAA2
UniProt ID: (Human) P54646, (Rat) Q09137, (Mouse) Q8BRK8
Entrez Gene ID: (Human) 5563, (Rat) 78975, (Mouse) 108079
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.