Invitrogen
Manufacturer Code:PA5114847
Catalog # PIPA5114847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AMPK alpha corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310008I11Rik 5'-AMP-activated protein kinase alpha-1 catalytic subunit 5'-AMP-activated protein kinase alpha-2 catalytic subunit 5'-AMP-activated protein kinase catalytic alpha-2 subunit 5'-AMP-activated protein kinase catalytic subunit alpha-1 5'-AMP-activated protein kinase catalytic subunit alpha-2 5'-AMP-activated protein kinase catalytic alpha-1 chain 5'-AMP-activated protein kinase catalytic alpha-2 chain 63 kDa subunit A830082D05 AAPK2 ACACA kinase acetyl-CoA carboxylase kinase AI194361 AI450832 AL024255 AMP -activate kinase alpha 1 subunit AMP-activated protein kinase AMP-activated protein kinase alpha 2 AMP-activated protein kinase alpha-2 subunit variant 2 AMP-activated protein kinase alpha-2 subunit variant 3 AMP-activated protein kinase alpha-2 variant B AMP-activated protein kinase alpha 1 catalytic subunit AMP-activated protein kinase catalytic alpha-1 AMPK AMPK 63 kDa subunit AMPK alpha 1 AMPK alpha 2 AMPK alpha-1 chain AMPK alpha-2 chain AMPK subunit alpha-1 AMPK subunit alpha-2 AMPK1 AMPK2 AMPKa1 AMPKa2 AMPK-activated protein kinase alpha-2 subunit AMPKalpha1 AMPK-alpha-1 chain AMPKalpha2 AMPK-alpha-2 chain C130083N04Rik HMGCR kinase hydroxymethylglutaryl-CoA reductase kinase pAMPK PRKAA Prkaa1 Prkaa2 protein kinase AMP-activated alpha 2 catalytic subunit protein kinase AMP-activated catalytic subunit alpha 1 protein kinase AMP-activated catalytic subunit alpha 2 protein kinase AMP-activated alpha 1 catalytic subunit protein kinase AMP-activated alpha 2 catalytic subunit tau-protein kinase PRKAA1; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK alpha 1; AMPK alpha 2; AMPK alpha-1 chain; AMPK alpha-2 chain; AMPK subunit alpha-1; AMPK subunit alpha-2; AMPK-alpha-2 chain; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; prkaa2; protein kinase, AMP-activated, alpha 1 catalytic subunit; protein kinase, AMP-activated, alpha 2 catalytic subunit; Tau-protein kinase PRKAA1
Gene Aliases: 2310008I11Rik; A830082D05; AI194361; AI450832; AL024255; AMPK; AMPK1; AMPK2; AMPKa1; AMPKa2; AMPKalpha1; AMPKalpha2; C130083N04Rik; PRKAA; PRKAA1; PRKAA2
UniProt ID: (Human) Q13131, (Human) P54646, (Rat) P54645, (Mouse) Q5EG47, (Rat) Q09137, (Mouse) Q8BRK8
Entrez Gene ID: (Human) 5562, (Human) 5563, (Rat) 65248, (Mouse) 105787, (Rat) 78975, (Mouse) 108079
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.