Invitrogen
Manufacturer Code:PA517398
Catalog # PIPA517398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Leu519 near the carboxy terminus of human AMPKa1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-AMP-activated protein kinase catalytic subunit alpha-1 5&prime-AMP-activated protein kinase catalytic alpha-1 chain AMP -activate kinase alpha 1 subunit AMP-activated protein kinase catalytic alpha-1 AMPK alpha 1 AMPK subunit alpha-1; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 63 kDa subunit; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK 63 kDa subunit; AMPK alpha 1; AMPK alpha-1 chain; AMPK subunit alpha-1; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; protein kinase, AMP-activated, alpha 1 catalytic subunit; Tau-protein kinase PRKAA1
Gene Aliases: AMPK; AMPK1; AMPKa1; PRKAA1
UniProt ID: (Human) Q13131
Entrez Gene ID: (Pig) 100145903, (Human) 5562
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.