Novus Biologicals
Manufacturer Code:NBP198543
Catalog # NBP198543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is AMPD3 - C-terminal region. Peptide sequence LQSGLSHQEKQKFLGQNYYKEGPEGNDIRKTNVAQIRMAFRYETLCNELS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase (isoform E); adenosine monophosphate deaminase (isoform E) adenosine monophosphate deaminase 3 AMP aminohydrolase AMP deaminase 3 AMP deaminase isoform E EC 3.5.4.6 Erythrocyte AMP deaminase erythrocyte type AMP deaminase erythrocyte-specific AMP deaminase myoadenylate deaminase; AMP aminohydrolase; AMP deaminase 3; AMP deaminase isoform E; Erythrocyte AMP deaminase; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; myoadenylate deaminase
Gene Aliases: AMPD3
UniProt ID: (Human) Q01432
Entrez Gene ID: (Human) 272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.