Abnova
Manufacturer Code:H00000271M09
Catalog # 89-016-142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6A8 |
Immunogen | AMPD2 (NP_631895 86 a.a. ~ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant AMPD2.
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 2 (isoform L); AMP deaminase 2; AMP deaminase isoform L; AMPD
Gene Aliases: AMPD2; PCH9; SPG63
UniProt ID: (Human) Q01433
Entrez Gene ID: (Human) 271
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.