Invitrogen
Manufacturer Code:PA529518
Catalog # PIPA529518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 429 and 774 of Human AMPD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 2 (isoform L); AMP deaminase 2; AMP deaminase 2 AMPD adenosine monophosphate deaminase 2 (isoform L); AMP deaminase isoform L; AMPD
Gene Aliases: AMPD2; PCH9; SPG63
UniProt ID: (Human) Q01433
Entrez Gene ID: (Human) 271
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.