Proteintech
Manufacturer Code:157101AP150UL
Catalog # 50-173-0423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AMPD2 Fusion Protein Ag8335 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 2 (isoform L); AMP deaminase 2; AMP deaminase 2 AMP deaminase isoform L AMPD2; AMP deaminase isoform L; AMPD
Gene Aliases: 1200014F01Rik; AI552571; Ampd; Ampd-2; AMPD2; PCH9; SPG63
UniProt ID: (Human) Q01433, (Mouse) Q9DBT5, (Rat) Q02356
Entrez Gene ID: (Human) 271, (Mouse) 109674, (Rat) 362015
Molecular Function: deaminase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.