Invitrogen
Manufacturer Code:PA555761
Catalog # PIPA555761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human AMPD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 1 (isoform M); adenosine monophosphate deaminase-1 (muscle); AMP deaminase 1; AMP deaminase isoform M; AMPD; Myoadenylate deaminase; RP5-1000E10.1 MAD MADA AMP deaminase 1 AMPD adenosine monophosphate deaminase 1 (isoform M) adenosine monophosphate deaminase-1 (muscle) myoadenylate deaminase skeletal muscle AMPD; skeletal muscle AMPD
Gene Aliases: AMPD1; MAD; MADA; MMDD
UniProt ID: (Human) P23109
Entrez Gene ID: (Human) 270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.