Invitrogen
Manufacturer Code:PA523172
Catalog # PIPA523172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (145 CIREKYMQKSFQRFPKTPSK 164) of AMPDA1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 1 (isoform M); adenosine monophosphate deaminase-1 (muscle); AMP deaminase 1; AMP deaminase isoform M; AMPD; Myoadenylate deaminase; RP5-1000E10.1 MAD MADA AMP deaminase 1 AMPD adenosine monophosphate deaminase 1 (isoform M) adenosine monophosphate deaminase-1 (muscle) myoadenylate deaminase skeletal muscle AMPD; skeletal muscle AMPD
Gene Aliases: AI553520; Ampd-1; Ampd01; AMPD1; MAD; MADA; MMDD; RATAMPD01
UniProt ID: (Human) P23109, (Rat) P10759, (Mouse) Q3V1D3
Entrez Gene ID: (Bovine) 512748, (Human) 270, (Dog) 606901, (Rat) 25028, (Mouse) 229665
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.